missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADPRM Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35281-100ul
This item is not returnable.
View return policy
Description
ADPRM Polyclonal antibody specifically detects ADPRM in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| ADPRM | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| ADPRibase-Mn, ADP-ribose/CDP-alcohol diphosphatase, manganese-dependent, ADPRM, C17orf48, chromosome 17 open reading frame 48, manganese-dependent ADP-ribose/CDP-alcohol diphosphatase, MDS006, NBLA03831 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 203-342 of human ADPRM (NP_064618.3).,, Sequence:, SEPQFVQFNGGFSQEQLNWLNEVLTFSDTNQEKVVIVSHLPIYPDASDNVCLAWNYRDALAVIWSHECVVCFFAGHTHDGGYSEDPFGVYHVNLEGVIETAPDSQAFGTVHVYPDKMMLKGRGRVPDRIMNYKKERAFHC | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 56985 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction