missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADPRHL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545851
This item is not returnable.
View return policy
Description
Peptide sequence: SVLDARELGM EERPYSSRLK KIGELLDQAS VTREEVVSEL GNGIAAFESV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
ADPRHL2 is a member of the ADP-ribosylglycohydrolase family. The enzyme catalyzes the removal of ADP-ribose from ADP-ribosylated proteins. This enzyme localizes to the mitochondria, in addition to the nucleus and cytoplasm.
Specifications
| ADPRHL2 | |
| Polyclonal | |
| Unconjugated | |
| ADPRHL2 | |
| [Protein ADP-ribosylarginine] hydrolase-like protein 2; [Protein ADP-ribosylserine] hydrolase; ADPRHL2; ADP-ribose glycohydrolase ARH3; ADP-ribose glycohydrolase ARH3; poly(ADP-ribose) glycohydrolase ARH3; ADP-ribosylarginine hydrolase like 2; ADP-ribosylhydrolase 2; ADP-ribosylhydrolase 3; ADP-ribosylhydrolase like 2; ADP-ribosylserine hydrolase; ADPRS; AI836109; ARH3; ecto ADP-ribosylhydrolase; LOW QUALITY PROTEIN: poly(ADP-ribose) glycohydrolase ARH3; O-acetyl-ADP-ribose deacetylase ARH3; Poly(ADP-ribose) glycohydrolase ARH3; protein ADP-ribosylarginine hydrolase-like protein 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 54936 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NX46 | |
| ADPRHL2 | |
| Synthetic peptide directed towards the C-terminal of human ADPRHL2 (aa 228-277). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction