missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADPRHL1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92248-0.02ml
This item is not returnable.
View return policy
Description
ADPRHL1 Polyclonal antibody specifically detects ADPRHL1 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| ADPRHL1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| [Protein ADP-ribosylarginine] hydrolase-like protein 1, ADP-ribosyl-hydrolase, ADP-ribosylhydrolase like 1, ARH2ADP-ribosylhydrolase 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 80-280 of human ADPRHL1 (NP_612439.2). YREMVRCYVEIVEKLPERRPDPATIEGCAQLKPNNYLLAWHTPFNEKGSGFGAATKAMCIGLRYWKPERLETLIEVSVECGRMTHNHPTGFLGSLCTALFVSFAAQGKPLVQWGRDMLRAVPLAEEYCRKTIRHTAEYQEHWFYFEAKWQFYLEERKISKDSENKAIFPDNYDAEEREKTYRKWSSEGRGGRRGHDAPMIA | |
| 0.02 mL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 113622 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction