missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADPGK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566176
This item is not returnable.
View return policy
Description
Immunogen sequence: SPGNGKDHSIL HSRNDLEEAF IHFMGKGAAA ERFFSDKETF HDIAQVASEF PGAQHYVGGN AALIGQKFAA NSDLKVLLC Highest antigen sequence identity to the following orthologs - mouse 90%, rat 89%.
The ADPGK gene encodes ADP-dependent glucokinase, an enzyme distinct from the ATP-dependent hexokinases as it employs adenosine diphosphate (ADP) rather than ATP to phosphorylate glucose to glucose-6-phosphate. This gene, initially identified in archaeal species, has been co-opted into mammalian systems where it plays roles primarily in glycolytic processes. Structurally, the mammalian ADPGK differs from its archaeal counterpart, exemplified by the rearrangement of its secondary structures, such as the alpha2 and alpha3 helices. These helices are replaced by an extended beta-strand region, indicating evolutionary adaptations for function in higher organisms like mice. Although its precise physiological role remains partially defined, ADPGK is notably expressed in both hematopoietic lineages and tumor tissues, suggesting potential significance in cancer biology and energy metabolism.
Specifications
| ADPGK | |
| Polyclonal | |
| Unconjugated | |
| ADPGK | |
| 2610017G09Rik; ADP dependent glucokinase; ADP-dependent glucokinase; ADPGK; Adp-gk; ATP-dependent glucokinase; C77577; EC 2.7.1.147; PSEC0260; RbBP-35; RGD1306103 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83440 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BRR6 | |
| ADPGK | |
| Recombinant Human ADPGK. Recombinant protein control fragment (Product #RP-106863). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction