Learn More
ADP-Sugar Pyrophosphatase/NUDT5 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Product Code. 18406859 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
Product Code. unitSize
18406859 0.1mL
1 options
Product Code. 18406859 Supplier Novus Biologicals Supplier No. NBP300064MFV500
Rabbit Polyclonal Antibody
ADP-Sugar Pyrophosphatase/NUDT5 Polyclonal antibody specifically detects ADP-Sugar Pyrophosphatase/NUDT5 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human ADP-Sugar Pyrophosphatase/NUDT5 (Uniprot#: Q9UKK9; Glu2-Phe219).