missing translation for 'onlineSavingsMsg'
Learn More

ADP-Sugar Pyrophosphatase/NUDT5 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18324953 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324953 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324953 Supplier Novus Biologicals Supplier No. NBP300064UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ADP-Sugar Pyrophosphatase/NUDT5 Polyclonal specifically detects ADP-Sugar Pyrophosphatase/NUDT5 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADP-Sugar Pyrophosphatase/NUDT5
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 3.6.1.-, EC 3.6.1.13, hYSAH1, nucleoside diphosphate linked moiety X-type motif 5, Nucleoside diphosphate-linked moiety X motif 5, nudix (nucleoside diphosphate linked moiety X)-type motif 5, Nudix motif 5, YSA1, YSA1HADP-sugar pyrophosphatase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ADP-Sugar Pyrophosphatase/NUDT5 (Q9UKK9; Glu2-Phe219).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 11164
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.