missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADP-ribosylarginine hydrolase Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-20168PE
This item is not returnable.
View return policy
Description
ADP-ribosylarginine hydrolase Polyclonal antibody specifically detects ADP-ribosylarginine hydrolase in Human samples. It is validated for ELISA
Specifications
| ADP-ribosylarginine hydrolase | |
| Polyclonal | |
| ELISA | |
| [Protein ADP-ribosylarginine] hydrolase, ADP-ribosylarginine hydrolaseARH1ADP-ribose-L-arginine cleaving enzyme, EC 3.2.2.19 | |
| Produced in rabbits immunized with purified, recombinant Human ADP-ribosylarginine hydrolase (Accession#: NP_001116.1; Met1-Leu351). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 141 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction