missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford ADP
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
CAS Number: 16178-48-6
Molecular Formula: C10H13N5O10P2Na2 2H2O
Molecular Weight: 507.2Specifications
Specifications
| CAS | 16178-48-6 |
| Molecular Formula | C10H13N5O10P2Na2-2H2O |
| Quantity | 10 g |
| Molecular Weight (g/mol) | 507.2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction