missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADORA2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572850
This item is not returnable.
View return policy
Description
ADORA2B Polyclonal Antibody for Western Blot, IHC (P), ELISA
ADORA2B encodes an adenosine receptor that is a member of the G protein-coupled receptor superfamily. This integral membrane protein stimulates adenylate cyclase activity in the presence of adenosine. This protein also interacts with netrin-1, which is involved in axon elongation. The gene is located near the Smith-Magenis syndrome region on chromosome 17.
Specifications
| ADORA2B | |
| Polyclonal | |
| Unconjugated | |
| ADORA2B | |
| A2b; A2b receptor; A2b, Rs; A2BAR; A2BR; AA2BR; adenosine A2B receptor; adenosine receptor A2b; ADORA2; Adora2b; AI480866; AI605384; HGNC:264; netrin-1 receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11541, 136, 29316 | |
| 4°C | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P29275, P29276, Q60614 | |
| ADORA2B | |
| A 19 amino acid peptide near the carboxy terminus of human ADORA2B, located within the last 50 amino acids. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction