missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADORA2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521305
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, rat liver. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ADORA2A is a protein which is one of several receptor subtypes for adenosine. The activity of the encoded protein, a G-protein coupled receptor family member, is mediated by G proteins which activate adenylyl cyclase. This protein is abundant in basal ganglia, vasculature and platelets and it is a major target of caffeine.
Specifications
| ADORA2A | |
| Polyclonal | |
| Unconjugated | |
| ADORA2A | |
| A2a receptor; A2AAR; A2aR; AA2AR; ADENO; adenosine A2a receptor; Adenosine A2a-receptor; adenosine receptor A2a; adenosine receptor subtype A2a; ADORA2; ADORA2A; Adora2l1; AR; hA2aR; RDC8; unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 135, 25369 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P29274, P30543 | |
| ADORA2A | |
| Synthetic peptide corresponding to a region within amino acids 255 and 345 of Adenosine A2a Receptor | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction