missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADORA2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533323
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human (100%); Gorilla, Gibbon, Monkey, Marmoset (95%); Dog (80%). Antigen retrieval requires Proteinase K treatment.
ADORA2A is a protein which is one of several receptor subtypes for adenosine. The activity of the encoded protein, a G-protein coupled receptor family member, is mediated by G proteins which activate adenylyl cyclase. This protein is abundant in basal ganglia, vasculature and platelets and it is a major target of caffeine.
Specifications
| ADORA2A | |
| Polyclonal | |
| Unconjugated | |
| ADORA2A | |
| A2a receptor; A2AAR; A2aR; AA2AR; ADENO; adenosine A2a receptor; Adenosine A2a-receptor; adenosine receptor A2a; adenosine receptor subtype A2a; ADORA2; ADORA2A; Adora2l1; AR; hA2aR; RDC8; unnamed protein product | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 135 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P29274 | |
| ADORA2A | |
| Synthetic 20 amino acid peptide from C-terminal cytoplasmic domain of human ADORA2A. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction