missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADORA2A 293T Cell Overexpression Lysate 1 (Denatured), Abnova
Description
Western Blotting
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | ADORA2A 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | NM_000675 |
| Molecular Weight (g/mol) | 45.43 |
| pH Range | 6.8 |
| Plasmid | pCMV-ADORA2A full-length |
| Quality Control Testing | Transient overexpression cell Lysate was tested with Anti-ADORA2A antibody (H00000135-B02) by western blotting |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Show More |
Safety and Handling
Product Identifier
- 100UL ADORA2A 293T Cell Transient Overexpression L
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contain: Mercaptoethanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?