missing translation for 'onlineSavingsMsg'
Learn More

Adonitol, Thermo Scientific™

Product Code. 15855058 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15855058 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15855058 Supplier Thermo Scientific Supplier No. J10635.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 488-81-3
Melting Point 101°C to 105°C
Packaging Plastic bottle
Molecular Formula C5H12O5
MDL Number MFCD00064291
Quantity 100 g
Beilstein 1720524
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
InChI Key HEBKCHPVOIAQTA-NGQZWQHPSA-N
SMILES C(C(C(C(CO)O)O)O)O
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
Molecular Weight (g/mol) 152.146
PubChem CID 6912
Formula Weight 152.15
Physical Form Powder
Chemical Name or Material Adonitol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.