missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adonitol, 99%

Product Code. 11322076
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322076 5 g 5g
11332076 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322076 Supplier Thermo Scientific Chemicals Supplier No. L03253.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 488-81-3 | C5H12O5 | 152.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Adonitol is metabolized to teicholic acids used in the cell walls of gram positive bacteria. Adonitol is often compared to other cell permeating molecules such as formamide, propanediol, and DMSO as a cryopreservation agent.

Solubility
Soluble in water (100 mg/ml).

Notes
Hygroscopic. Store under dry inert gas. Incompatible with water and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-81-3
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.146
MDL Number MFCD00064291
InChI Key HEBKCHPVOIAQTA-NGQZWQHPSA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
SMILES C(C(C(C(CO)O)O)O)O

Specifications

Melting Point 102°C to 104°C
Quantity 5 g
Beilstein 1720524
Sensitivity Hygroscopic
Merck Index 14,168
Solubility Information Soluble in water (100mg/ml).
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material Adonitol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.