missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adonitol, 98%

Product Code. 10667332 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
10g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10667332 10 g Glass bottle 10g
10657332 5 g Glass bottle 5g
10793085 100 g Glass bottle 100g
11904641 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10667332 Supplier Thermo Scientific Chemicals Supplier No. 167030100
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Adonitol, 98%, C5H12O5, CAS Number-488-81-3, adonit, adonite, adonitol, d-ribitol, d-xylitol, eutrit, ribitol, xylit, xylite, xylitol, 100g, 97.5% min. (ELSD) (HPLC), 01,53, 100.0, 99.0, White, 207-685-7, 152.15, HEBKCHPVOIAQTA-NGQZWQHPSA-N, Authentic, (2S,4R)-pentane-1,2,3,4,5-pentol | CAS: 488-81-3 | C5H12O5 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-81-3
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064291
InChI Key HEBKCHPVOIAQTA-NGQZWQHPSA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
SMILES C(C(C(C(CO)O)O)O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 101.0°C to 104.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 01,53
Merck Index 15,157
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hot alcohol,insoluble in ether
Formula Weight 152.15
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Adonitol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.