missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AdoHcyase 3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562735
This item is not returnable.
View return policy
Description
Immunogen sequence: GKVPQASAMK RSDPHHQHQR HRDGGEALVS PDGTVTEAPR TVKKQIQFAD QKQEFNKRPT KI Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 98%.
The protein encoded by this gene acts as a homotetramer and may be involved in the conversion of S-adenosyl-L-homocysteine to L-homocysteine and adenosine. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| AdoHcyase 3 | |
| Polyclonal | |
| Unconjugated | |
| AHCYL2 | |
| Adenosylhomocysteinase 3; adenosylhomocysteinase like 2; adenosylhomocysteinase-like 2; AdoHcyase 3; ADOHCYASE3; AHCYL2; IP(3)Rs binding protein released with IP(3) 2; IRBIT2; KIAA0828; Long-IRBIT; putative adenosylhomocysteinase 3; S-adenosylhomocysteine hydrolase-like 2; S-adenosylhomocysteine hydrolase-like protein 2; S-adenosyl-L-homocysteine hydrolase 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23382 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96HN2 | |
| AHCYL2 | |
| Recombinant protein corresponding to Human AdoHcyase 3. Recombinant protein control fragment (Product #RP-98530). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction