missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AdoHcyase 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560038
This item is not returnable.
View return policy
Description
Immunogen sequence: MSMPDAMPLP GVGEELKQAK EIEDAEKYSF MATVTKAPKK QIQFADDMQE FTK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 30%.
The protein encoded by this gene interacts with inositol 1,4,5-trisphosphate receptor, type 1 and may be involved in the conversion of S-adenosyl-L-homocysteine to L-homocysteine and adenosine. Several transcript variants encoding two different isoforms have been found for this gene.
Specifications
| AdoHcyase 2 | |
| Polyclonal | |
| Unconjugated | |
| AHCYL1 | |
| 1110034F20Rik; AA409031; AA414901; adenosylhomocysteinase 2; adenosylhomocysteinase 2; S-adenosylhomocysteine hydrolase-like protein 1; adenosylhomocysteinase like 1; adenosylhomocysteinase-like 1; adoHcyase 2; Ahcyl1; Ahcyl1-v1; Ahcyl1-v2; Ahcy-rs3; cb586; DCAL; DC-expressed AHCY-like molecule; dendritic cell expressed AHCY-like protein; inositol 1,4,5-trisphosphate receptor-binding protein; IP(3)Rs binding protein released with IP(3); IP3R binding protein released with inositol 1,4,5-trisphosphate; IP3R-binding protein released with inositol 1,4,5-trisphosphate; Irbit; PPP1R78; PRO0233; protein phosphatase 1, regulatory subunit 78; putative adenosylhomocysteinase 2; s adenosylhomocysteine hydrolase; S-adenosyl homocysteine hydrolase homolog; S-adenosylhomocysteine hydrolase, related sequence 3; S-adenosylhomocysteine hydrolase-like 1; S-adenosylhomocysteine hydrolase-like protein 1; S-adenosylhomocysteine hydrolase-like protein variant 1; S-adenosylhomocysteine hydrolase-like protein variant 2; S-adenosyl-L-homocysteine hydrolase 2; wu:fj66f02; XPVKONA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10768, 229709, 362013 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| B5DFN2, O43865, Q80SW1 | |
| AHCYL1 | |
| Recombinant protein corresponding to Human AdoHcyase 2. Recombinant protein control fragment (Product #RP-99810). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction