missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADO Polyclonal Antibody
GREENER_CHOICE

Product Code. 15964315
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15964315 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15964315 Supplier Invitrogen™ Supplier No. PA578733

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human PC-3 whole cell, human MCF-7 whole cell, rat brain tissue, mouse heart tissue, mouse brain tissue. IHC: rat brain tissue, mouse brain tissue. ICC/IF: A549 cell. Flow: A549 cell, PC-3 cell, U251 cell, Hela cell.

Human thiol dioxygenases include cysteine dioxygenase (CDO; MIM 603943) and cysteamine (2-amino ethane thiol) dioxygenase (ADO; EC 1.13.11.19). CDO adds 2 oxygen atoms to free cysteine, whereas ADO adds 2 oxygen atoms to free cysteamine to form hypotaurine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADO
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene ADO
Gene Accession No. Q6PDY2, Q96SZ5
Gene Alias 2-aminoethanethiol (cysteamine) dioxygenase; 2-aminoethanethiol dioxygenase; ADO; C10orf22; cysteamine (2-aminoethanethiol) dioxygenase (ADO); cysteamine dioxygenase; Gm237
Gene Symbols ADO
Host Species Rabbit
Immunogen E. coli-derived human ADO recombinant protein (Position: E49-E261).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 211488, 309732, 84890
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.