Learn More
Invitrogen™ ADNP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552286
Description
Immunogen sequence: SGSPFDPVFE VEPKISNDNP EEHVLKVIPE DASESEEKLD QKEDGSKYET IHLTEEPTKL MHNASDSEVD QDDVVEWKDG ASPSESGPGS QQVSDFEDNT CEMKPGTWSD ESSQSEDARS SKPAAKKKAT MQGDREQLKW KNSSYGKVEG Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.
Vasoactive intestinal peptide is a neuroprotective factor that has a stimulatory effect on the growth of some tumor cells and an inhibitory effect on others. This gene encodes a protein that is upregulated by vasoactive intestinal peptide and may be involved in its stimulatory effect on certain tumor cells. The encoded protein contains one homeobox and nine zinc finger domains, suggesting that it functions as a transcription factor. This gene is also upregulated in normal proliferative tissues. Finally, the encoded protein may increase the viability of certain cell types through modulation of p53 activity. Alternatively spliced transcript variants encoding the same protein have been described.
Specifications
| ADNP | |
| Polyclonal | |
| Unconjugated | |
| Adnp | |
| AA589558; activity dependent neuroprotector homeobox; activity-dependent neuroprotective protein; activity-dependent neuroprotector; activity-dependent neuroprotector homeobox; activity-dependent neuroprotector homeobox protein; Adnp; ADNP homeobox 1; ADNP1; HVDAS; KIAA0784; mKIAA0784; MRD28; NAP peptide | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11538, 23394, 64622 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H2P0, Q9JKL8, Q9Z103 | |
| Adnp | |
| Recombinant protein corresponding to Human ADNP. Recombinant protein control fragment (Product #RP-102484). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.