missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADM2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550482
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total ADM2 protein.
ADM2 Polyclonal antibody specifically detects ADM2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
ADM2 | |
Polyclonal | |
Unconjugated | |
ADM2 | |
ADM2; adrenomedullin 2; Adrenomedullin-2; AM2; dJ579N16.4; Imd; IMDL; Imdn; IMDS; Intermedin; Intermedin-long; Intermedin-short; Protein ADM2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79924 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q7Z4H4 | |
ADM2 | |
Synthetic peptide corresponding to a region derived from internal residues of human adrenomedullin 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |