missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADK Rabbit anti-Human, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90184-50UL
This item is not returnable.
View return policy
Description
ADK Monoclonal specifically detects ADK in Human samples. It is validated for ELISA.
Specifications
| ADK | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 1 | |
| ELISA 1:5000-1:10000 | |
| adenosine kinase, AKAdenosine 5′-phosphotransferase, EC 2.7.1, EC 2.7.1.20 | |
| Recombinant Human ADK protein | |
| 50 μg | |
| Cancer, Cell Cycle and Replication, Immunology, Lipid and Metabolism | |
| 132 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction