missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADK, Mouse anti-Human, Clone: AT4F8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB16188.50uL
This item is not returnable.
View return policy
Specifications
| ADK | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.4 (0.09% sodium azide). | |
| AK | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| AT4F8 | |
| ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| adenosine kinase | |
| ADK | |
| Recombinant protein corresponding to human ADK. | |
| RUO | |
| 132 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction