Learn More
Invitrogen™ ADK Monoclonal Antibody (7F4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549220
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Human ADK shares 88.8% and 88.2% amino acid (aa) sequence identity with mouse and rat ADK, respectively. Positive Control - WB: human PC-3 whole cell, human U-87MG whole cell, human SW620 whole cell. IHC: human placenta tissue, human tonsil tissue, human rectal cancer tissue. Flow: PC-3 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The protein encoded by this gene catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| ADK | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P55263 | |
| ADK | |
| E. coli-derived human ADK recombinant protein (Position: K165-T351). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 7F4 | |
| Unconjugated | |
| ADK | |
| 2310026J05Rik; 5033405D03Rik; Adenosine 5'-phosphotransferase; adenosine kinase; ADK; AI255373; AI987814; AK; testicular tissue protein Li 14 | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 132 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.