Learn More
Yes 2 Solutions™ Adjustable set square with bevel and graduated arch
Product Code. 15424314
Encompass_Preferred
Product Code. 15424314 Supplier Yes 2 Solutions™ Supplier No. LX12011
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured in one part with the arm which is precision divided in half degrees, tangent and secant. Manufactured from clear glass Dunilon.
250mm