Learn More
Erlab Adjustable Base Without Wheels - Loïs 1500
Product Code. 17842664
Click to view available options
Product Type:
Laminar Flow Cabinet
Unit Size:
Each
Product Code. 17842664
Brand: Erlab NORT000409
Adapted base for your Laminar Airflow Cabinet.
  • Without wheels
  • Without central leg
  • Compatible model: PCR cabinet Loïs 1500
Specifications
Product Type Laminar Flow Cabinet