missing translation for 'onlineSavingsMsg'
Learn More

Adipoyl Chloride, TRC

Product Code. 30326761
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326761 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326761 Supplier Toronto Research Chemicals Supplier No. TRCA291700100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Adipoyl Chloride
CAS 111-50-2
Molecular Formula C6 H8 Cl2 O2
Quantity 100 g
Synonym Hexanedioyl Dichloride, 1,4-Bis(chlorocarbonyl)butane, Adipic Acid Dichloride, Adipoyl Dichloride, Adipyl Chloride, Adipyl Dichloride, Hexanedioic Acid Dichloride, Hexanedioic Dichloride
InChI Formula InChI=1S/C6H8Cl2O2/c7-5(9)3-1-2-4-6(8)10/h1-4H2
SMILES ClC(=O)CCCCC(=O)Cl
IUPAC Name hexanedioyl dichloride
Molecular Weight (g/mol) 183.03
Formula Weight 181.9901
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.