missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ AdipoRed™ Assay Reagent

Product Code. 11640381
Change view
Click to view available options
Quantity:
5 x 4 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11640381 5 x 4 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11640381 Supplier Lonza™ Supplier No. PT7009
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Lonza™ AdipoRed™ Assay Reagent is designed for assessing the effect of compounds on the differentiation of preadipocytes or on lipid utilization in mature adipocytes. The lipophilic reagent partitions into the fat droplets of differentiated adipocytes and fluoresces at 572nm.

AdipoRed™ Assay Reagent is used to quantify intracellular lipid accumulation. This objective, high-throughput, homogeneous fluorescence-based assay quantifies the accumulation of intracellular triglycerides and provides significant advantages to drug discovery efforts in the field of obesity and diabetes. It is more sensitive than other methods such as the Oil Red O assay, and is much faster and easier than Northern and Western blots.

  • Convenient: Simply remove cell culture medium, add premixed reagent and read in a standard fluorometer
  • Fast: Process an entire 96-well or 384-well plate in as little as 20 minutes
  • Effective: Provides objective measurement of the accumulation of intracellular triglycerides, with high signal to noise ratios
  • Easy: Packaged in five premeasured, single use 4mL bottles
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 189°C
CAS 67-68-5
Detection Standard fluorometer
Color Red
Form Liquid
Formulation Dimethyl sulfoxide 99.99% w/w
For Use With (Application) Used to quantify intracellular lipid accumulation
Kit Contents 5 premixed reagent
Melting Temperature 18.4°C
Odor Odorless
Quantity 5 x 4 mL
Range Fluoresces at 572nm
Stained Intracellular triglycerides
Storage Requirements Store at -20°C
Product Type Adipogenesis Assay Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.