missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AdipoR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30867-25ug
This item is not returnable.
View return policy
Description
AdipoR2 Polyclonal antibody specifically detects AdipoR2 in Human samples. It is validated for ELISA,Western Blot
Specifications
| AdipoR2 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL, ELISA | |
| ACDCR2, adiponectin receptor 2, adiponectin receptor protein 2, FLJ21432, PAQR2MGC4640, Progestin and adipoQ receptor family member II | |
| Rabbits were immunized with a synthetic peptide near the C terminus of human AdipoR2 proteins | |
| 25 μg | |
| Signal Transduction | |
| 79602 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| ELISA, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction