missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADIPOQ Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21613-1-AP-150UL
This item is not returnable.
View return policy
Description
Adiponectin is a protein hormone secreted exclusively by adipocytes that regulates the metabolism of lipids and glucose. Adiponectin has been shown to play a role in various physiological processes including homeostasis and obesity. Decreased levels of adiponectin are associated with obese individuals as well as insulin resistance and hyperinsulinemia (type 2 diabetes). In mice adiponectin has been shown to improve insulin signaling, thus ameliorating insulin resistance. Through activation of AMP kinase and PPAR-alpha, adiponectin can increase fatty acid transport, oxidation, and dissipation in skeletal muscle, leading to a reduction of intramyocellular lipid levels and thus improved insulin signaling.Specifications
| ADIPOQ | |
| Polyclonal | |
| Unconjugated | |
| ADIPOQ | |
| ACDC, ACRP30, adiponectin, ADIPOQ, ADIPQTL1, ADPN, APM 1, APM1, GBP28, Gelatin binding protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11450, 9370 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15848, Q60994 | |
| ADIPOQ | |
| ADIPOQ Fusion Protein Ag16304 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction