missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Adiponutrin/PNPLA3 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
Description
A blocking peptide from PNPLA3. Source: Synthetic Amino Acid Sequence: (Accession #: NP_473429.2) EHDICPKVKSTN The Adiponutrin/PNPLA3 Blocking Peptide is derived from Synthetic. The Adiponutrin/PNPLA3 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 80339 |
| Species | Mouse |
| Purity | >50%, by HPLC |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Format | Lyophilized |
| Formulation | Lyophilized from a 5 pmol/ul solution in 5% (v/v) acetonitrile/H2O and TFA salt. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-00227. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | Acylglycerol O-acyltransferase, adiponutrin, ADPNdJ796I17.1, C22orf20, Calcium-independent phospholipase A2-epsilon, chromosome 22 open reading frame 20, EC 2.3.1.-, EC 2.7.7.56, EC 3.1.1.3, EC 4.2.3.4, FLJ22012, iPLA(2)epsilon, iPLA2-epsilon, patatin-like phospholipase domain containing 3, patatin-like phospholipase domain-containing protein 3 |
| Gene Symbol | PNPLA3 |
| Quantity | 0.1 mg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction