missing translation for 'onlineSavingsMsg'
Learn More

Adiponutrin/PNPLA3 Antibody [PE], Novus Biologicals™

Product Code. 30735985 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735985 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735985 Supplier Novus Biologicals Supplier No. AF5208P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Adiponutrin/PNPLA3 Polyclonal antibody specifically detects Adiponutrin/PNPLA3 in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adiponutrin/PNPLA3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias Acylglycerol O-acyltransferase, adiponutrin, ADPNdJ796I17.1, C22orf20, Calcium-independent phospholipase A2-epsilon, chromosome 22 open reading frame 20, EC 2.3.1.-, EC 2.7.7.56, EC 3.1.1.3, EC 4.2.3.4, FLJ22012, iPLA(2)epsilon, iPLA2-epsilon, patatin-like phospholipase domain containing 3, patatin-like phospholipase domain-containing protein 3
Host Species Sheep
Immunogen E. coli-derived recombinant human Adiponutrin/PNPLA3, Arg160-Arg349, Accession # Q9NST1
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 80339
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.