missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Adiponectin Receptor 2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5114166

Product Code. 17162024

  • £439.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: FPGKCDIWFH SHQLFHIFVV AGAFVHFHGV SNLQEFRFMI GGGCSEEDAL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Goat: 92%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%.

Adiponectin (also known as 30-kDa adipocyte complement-related protein; Acrp30) is a hormone secreted by adipocytes that acts as an antidiabetic and anti-atherogenic adipokine. Levels of adiponectin in the blood are decreased under conditions of obesity, insulin resistance and type 2 diabetes. Administration of adiponectin causes glucose-lowering effects and ameliorates insulin resistance in mice. Conversely, adiponectin-deficient mice exhibit insulin resistance and diabetes. This insulin-sensitizing effect of adiponectin seems to be mediated by an increase in fatty-acid oxidation through activation of AMP kinase and PPAR. Here we report the cloning of complementary DNAs encoding adiponectin receptors 1 and 2 (AdipoR1 and AdipoR2) by expression cloning. AdipoR1 is abundantly expressed in skeletal muscle, whereas AdipoR2 is predominantly expressed in the liver. These two adiponectin receptors are predicted to contain seven transmembrane domains, but are structurally and functionally distinct from G-protein-coupled receptors. Expression of AdipoR1/R2 or suppression of AdipoR1/R2 expression by small-interfering RNA supports our conclusion that they serve as receptors for globular and full-length adiponectin, and that they mediate increased AMP kinase and PPAR- ligand activities, as well as fatty-acid oxidation and glucose uptake by adiponectin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Adiponectin Receptor 2
Polyclonal
Unconjugated
ADIPOR2
1110001I14Rik; ACDCR2; ADCR2; adiponectin receptor 2; adiponectin receptor protein 2; Adipor2; AI115388; AW554121; D6Ucla1e; FLJ21432; MGC4640; Paqr2; Parq2; Progestin and adipoQ receptor family member 2; Progestin and adipoQ receptor family member II
Rabbit
Affinity chromatography
RUO
79602
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q86V24
ADIPOR2
Immunogen sequence: FPGKCDIWFH SHQLFHIFVV AGAFVHFHGV SNLQEFRFMI GGGCSEEDAL.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.