Learn More
Invitrogen™ Adiponectin Receptor 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1059
Description
PA1-059 detects human and rat adiponectin receptor 1. PA1-059 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~42 kDa protein corresponding to human adiponectin receptor 1 from transfected hepatocytes. The PA1-059 immunogen is a synthetic peptide corresponding to residues R(20) E A D T V E L A E L G P L L E E K(37) of human Adiponectin Receptor 1.
Adiponectin (also known as 30- kDa adipocyte complement-related protein; Acrp30) is a hormone secreted by adipocytes that acts as an antidiabetic and anti-atherogenic adipokine. Levels of adiponectin in the blood are decreased under conditions of obesity, insulin resistance and type 2 diabetes. Administration of adiponectin causes glucose-lowering effects and ameliorates insulin resistance in mice. Conversely, adiponectin-deficient mice exhibit insulin resistance and diabetes. This insulin-sensitizing effect of adiponectin seems to be mediated by an increase in fatty-acid oxidation through activation of AMP kinase and PPAR-. Here we report the cloning of complementary DNAs encoding adiponectin receptors 1 and 2 (AdipoR1 and AdipoR2) by expression cloning. AdipoR1 is abundantly expressed in skeletal muscle, whereas AdipoR2 is predominantly expressed in the liver. These two adiponectin receptors are predicted to contain seven transmembrane domains, but are structurally and functionally distinct from G-protein-coupled receptors. Expression of AdipoR1/R2 or suppression of AdipoR1/R2 expression by small-interfering RNA supports our conclusion that they serve as receptors for globular and full-length adiponectin, and that they mediate increased AMP kinase and PPAR- ligand activities, as well as fatty-acid oxidation and glucose uptake by adiponectin.
Specifications
| Adiponectin Receptor 1 | |
| Polyclonal | |
| Unconjugated | |
| ADIPOR1 | |
| 2810031L11Rik; ACDCR1; adiponectin receptor 1; adiponectin receptor protein 1; Adipor1; CGI45; CGI-45; PAQR1; Parq1; Progestin and adipoQ receptor family member 1; Progestin and adipoQ receptor family member I; TESBP1A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 289036, 51094 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| Q96A54 | |
| ADIPOR1 | |
| Synthetic peptide corresponding to residues R(20) E A D T V E L A E L G P L L E E K(37) of human Adiponectin Receptor 1. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.