missing translation for 'onlineSavingsMsg'
Learn More

Adiponectin/Acrp30 Antibody (166126) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30736691 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736691 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736691 Supplier Novus Biologicals Supplier No. FAB10651JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Adiponectin/Acrp30 Monoclonal antibody specifically detects Adiponectin/Acrp30 in Human samples. It is validated for ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adiponectin/Acrp30
Applications ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 166126
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias ACDC, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, APM1APM-1, C1q and collagen domain-containing protein, GBP28apM1, Gelatin-binding protein
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Adiponectin, Glu19-Asn244, Accession # Q15848
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 9370
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.