missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adipic dihydrazide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Adipic dihydrazide is used as a formaldehyde scavenger and reacts with formaldehyde, thereby preventing the volatilizing of formaldehyde in the air. It is also employed as a paint additive and coating additive. It is also used as an intermediate. Further, it is used for cross-linking water-based emulsions and as a hardener for certain epoxy resins, which finds application in powder coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Adipic dihydrazide is used as a formaldehyde scavenger and reacts with formaldehyde, thereby preventing the volatilizing of formaldehyde in the air. It is also employed as a paint additive and coating additive. It is also used as an intermediate. Further, it is used for cross-linking water-based emulsions and as a hardener for certain epoxy resins, which finds application in powder coating.
Solubility
Soluble in water, alcohol and acetone.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
Adipic dihydrazide is used as a formaldehyde scavenger and reacts with formaldehyde, thereby preventing the volatilizing of formaldehyde in the air. It is also employed as a paint additive and coating additive. It is also used as an intermediate. Further, it is used for cross-linking water-based emulsions and as a hardener for certain epoxy resins, which finds application in powder coating.
Solubility
Soluble in water, alcohol and acetone.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
Specifications
Melting Point | 180°C to 182°C |
Odor | Odorless |
Quantity | 100 g |
Beilstein | 973863 |
Solubility Information | Soluble in water,alcohol and acetone. |
Formula Weight | 174.2 |
Percent Purity | 97% |
Chemical Name or Material | Adipic dihydrazide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'