missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adipic Acid Monoethyl Ester, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Adipic Acid Monoethyl Ester |
| CAS | 626-86-8 |
| Molecular Formula | C8 H14 O4 |
| Quantity | 100 g |
| Synonym | Hexanedioic Acid Monoethyl Ester, 1,4-Butanedicarboxylic Acid Monoethyl Ester, 1,6-Hexandioic Acid Ethyl Ester, 6-Ethoxy-6-oxohexanoic Acid, Ethyl Hydrogen Adipate, Ethyl Hydrogen Hexanedioate, Monoethyl Adipate, Monoethyl Hexanedioate, NSC 8877 |
| InChI Formula | InChI=1S/C8H14O4/c1-2-12-8(11)6-4-3-5-7(9)10/h2-6H2,1H3,(H,9,10) |
| SMILES | CCOC(=O)CCCCC(=O)O |
| IUPAC Name | 6-ethoxy-6-oxohexanoic acid |
| Molecular Weight (g/mol) | 174.194 |
| Formula Weight | 174.089 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction