missing translation for 'onlineSavingsMsg'
Learn More

Adipic Acid Monoethyl Ester, TRC

Product Code. 30324633
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 30324633

Brand: Toronto Research Chemicals TRCA14489025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Adipic Acid Monoethyl Ester
CAS 626-86-8
Molecular Formula C8 H14 O4
Quantity 25 g
Synonym Hexanedioic Acid Monoethyl Ester, 1,4-Butanedicarboxylic Acid Monoethyl Ester, 1,6-Hexandioic Acid Ethyl Ester, 6-Ethoxy-6-oxohexanoic Acid, Ethyl Hydrogen Adipate, Ethyl Hydrogen Hexanedioate, Monoethyl Adipate, Monoethyl Hexanedioate, NSC 8877
InChI Formula InChI=1S/C8H14O4/c1-2-12-8(11)6-4-3-5-7(9)10/h2-6H2,1H3,(H,9,10)
SMILES CCOC(=O)CCCCC(=O)O
IUPAC Name 6-ethoxy-6-oxohexanoic acid
Molecular Weight (g/mol) 174.194
Formula Weight 174.089
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.