missing translation for 'onlineSavingsMsg'
Learn More

Adipic acid monoethyl ester, 95%

Product Code. 11486906
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11486906

Brand: Thermo Scientific Alfa Aesar B24006.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 626-86-8 | C8H14O4 | 174.196 g/mol

Adipic acid monoethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adipic acid monoethyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-86-8
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00004419
InChI Key UZNLHJCCGYKCIL-UHFFFAOYSA-N
Synonym adipic acid monoethyl ester, monoethyl adipate, ethyl hydrogen adipate, hexanedioic acid, monoethyl ester, monoethyl hexanedioate, monoethyladipic acid ester, adipic acid, monoethyl ester, mono-ethyl adipate, hexanedioic acid, 1-ethyl ester, hexanoic acid, monoethyl ester
PubChem CID 12295
IUPAC Name 6-ethoxy-6-oxohexanoic acid
SMILES CCOC(=O)CCCCC(=O)O

Specifications

Melting Point 28°C to 29°C
Density 0.98
Boiling Point 179°C to 181°C (decomposition)
Flash Point >110°C (230°F)
Refractive Index 1.439
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 174.2
Percent Purity 95%
Chemical Name or Material Adipic acid monoethyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.