missing translation for 'onlineSavingsMsg'
Learn More

Adipic Acid, Honeywell Fluka™

Product Code. p-7071955 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
25 kg
Unit Size:
100g
25kg
500g
This item is not returnable. View return policy

Product Code. 15622920

Brand: Honeywell-Fluka 27635100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Meets analytical specification of Ph.Eur., BP, E 355, 99.6-101.0% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-04-9
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00004420
InChI Key WNLRTRBMVRJNCN-UHFFFAOYSA-N
Synonym adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german
PubChem CID 196
ChEBI CHEBI:30832
IUPAC Name hexanedioic acid
SMILES C(CCC(=O)O)CC(=O)O

Specifications

Linear Formula HOOC(CH2)4COOH
Beilstein 1209788
Solubility Information Freely soluble in methanol, boiling water, cyclohexane, acetone, water, ethanol. Insoluble in benzene,petroleum ether.
Formula Weight 146.14g/mol
Percent Purity 99.6 to 101% (calc. to the dried substance)
Quantity 100 g
Flash Point 196°C (384.8°F)
Ignition Point 788°F
Residue after Ignition ≤0.002% (as SO4)
Vapor Density 5 (vs air)
Vapor Pressure 1mmHg (159.5°C)
Chemical Name or Material Adipic Acid, Meets analytical specification of Ph.Eur.BP
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.