missing translation for 'onlineSavingsMsg'
Learn More

Adipic Acid, Solstice

Product Code. 15642920 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
25 kg
Unit Size:
100g
25kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15642920 500 g 500g
15622920 100 g 100g
15632920 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15642920 Supplier Honeywell-Fluka Supplier No. 27635500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Meets analytical specification of Ph.Eur., BP, E 355, 99.6-101.0% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-04-9
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00004420
InChI Key WNLRTRBMVRJNCN-UHFFFAOYSA-N
Synonym adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german
PubChem CID 196
ChEBI CHEBI:30832
IUPAC Name hexanedioic acid
SMILES C(CCC(=O)O)CC(=O)O

Specifications

Linear Formula HOOC(CH2)4COOH
Beilstein 1209788
Solubility Information Freely soluble in methanol, boiling water, cyclohexane, acetone, water, ethanol. Insoluble in benzene,petroleum ether.
Formula Weight 146.14g/mol
Percent Purity 99.6 to 101% (calc. to the dried substance)
Quantity 500 g
Flash Point 196°C (384.8°F)
Ignition Point 788°F
Residue after Ignition ≤0.002% (as SO4)
Vapor Density 5 (vs air)
Vapor Pressure 1mmHg (159.5°C)
Chemical Name or Material Adipic Acid, Meets analytical specification of Ph.Eur.BP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.