missing translation for 'onlineSavingsMsg'
Learn More

Adipic acid, Dr. Ehrenstorfer

Product Code. 30321837
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321837 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321837 Supplier Dr. Ehrenstorfer Supplier No. DREC10045900100MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Adipic acid (Hexanedioic acid)
CAS 124-04-9
Analyte or Component Names Adipic acid (Hexanedioic acid)
Shipping Condition +20°C
Molecular Formula C6 H10 O4
Certifications/Compliance ISO 17034
Synonym Adipic acid, Hexanedioic acid
InChI Formula InChI=1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)
SMILES OC(=O)CCCCC(=O)O
IUPAC Name hexanedioic acid
Molecular Weight (g/mol) 146.14
Quantity 100 mg
Formula Weight 146.0579 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.