missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adipic acid, 99%, Thermo Scientific Chemicals
£23.10 - £259.00
Chemical Identifiers
CAS | 124-04-9 |
---|---|
Molecular Formula | C6H10O4 |
Molecular Weight (g/mol) | 146.142 |
MDL Number | MFCD00004420 |
InChI Key | WNLRTRBMVRJNCN-UHFFFAOYSA-N |
Synonym | adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german |
PubChem CID | 196 |
ChEBI | CHEBI:30832 |
IUPAC Name | hexanedioic acid |
SMILES | C(CCC(=O)O)CC(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10132320
|
Thermo Scientific Acros
102815000 |
500 g | Plastic bottle |
£23.10
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10090550
|
Thermo Scientific Acros
102810010 |
1 kg | Plastic bottle |
£29.90
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10538280
|
Thermo Scientific Acros
102810025 |
2.5 kg | Plastic bottle |
£51.10
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10454745
|
Thermo Scientific Acros
102810250 |
25 kg | Fibre drum |
£259.00
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
124-04-9 | |
146.142 | |
WNLRTRBMVRJNCN-UHFFFAOYSA-N | |
196 | |
hexanedioic acid |
C6H10O4 | |
MFCD00004420 | |
adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german | |
CHEBI:30832 | |
C(CCC(=O)O)CC(=O)O |
Specifications
124-04-9 | |
337.0°C | |
100.0 | |
99% | |
HO2C(CH2)4CO2H | |
02, 649 | |
15, 150 | |
WNLRTRBMVRJNCN-UHFFFAOYSA-N | |
hexanedioic acid | |
196 | |
146.14 | |
196°C | |
Crystalline Powder |
151.0°C to 153.0°C | |
98.5 | |
White | |
C6H10O4 | |
MFCD00004420 | |
01,15 | |
adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german | |
C(CCC(=O)O)CC(=O)O | |
146.142 | |
CHEBI:30832 | |
99% | |
Authentic | |
Adipic acid |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Adipic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only