Learn More
Invitrogen™ ADI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592300
Description
Immunogen sequence: MVQAWYMDDA PGDPRQPHRP DPGRPVGLEQ LRRLGVLYWK LDADKYENDP ELEKIRRERN YSWMDIITIC KDKLPNYEEK IKMFYEEHLH LDDEIRYILD GSGYFDVRDK EDQWIRIFME KGDMVTLPAG IYHRFTVDEK NYTKAMRLFV GEPVWTAYNR PADHFEARGQ YVKFLAQTA; Positive Samples: U-87MG, A-549, SGC7901, HT-29, 293T, HepG2; Cellular Location: Cell membrane, Cytoplasm, Cytoplasmic side, Nucleus, Peripheral membrane protein.
This gene encodes an enzyme that belongs to the aci-reductone dioxygenase family of metal-binding enzymes, which are involved in methionine salvage. This enzyme may regulate mRNA processing in the nucleus, and may carry out different functions depending on its localization. A pseudogene that is located on chromosome 20 has been defined for this gene.
Specifications
| ADI1 | |
| Polyclonal | |
| Unconjugated | |
| ADI1 | |
| 1,2-dihydroxy-3-keto-5-methylthiopentene dioxygenase; 1,2-dihydroxy-3-keto-5-methylthiopentene dioxygenase {ECO:0000255; aci-reductone dioxygenase; aci-reductone dioxygenase (ARD)-like protein 1; acireductone dioxygenase (Fe(2+)-requiring); acireductone dioxygenase (Fe(2+)-requiring) {ECO:0000255; acireductone dioxygenase (Ni(2+)-requiring); acireductone dioxygenase 1; ADI1; AL024210; Alp1; Androgen-responsive ARD-like protein 1; androgen-responsive gene encoding an ARD-like protein; APL1; ARD; ARD {ECO:0000255; fe-ARD; fe-ARD {ECO:0000255; HAMAP-Rule:MF_03154}; HMFT1638; membrane-type 1 matrix metalloproteinase cytoplasmic tail binding protein-1; membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1; membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1 {ECO:0000255; MT1-MMP cytoplasmic tail-binding protein-1; MTCBP1; MTCBP-1; MTCBP-1 {ECO:0000255; mtnD; Ni-ARD; SIPL; Sip-L; submergence induced protein 2; submergence-induced protein-like factor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 104923, 298934, 55256 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q562C9, Q99JT9, Q9BV57 | |
| ADI1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-179 of human ADI1 (NP_0607392). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.