missing translation for 'onlineSavingsMsg'
Learn More

ADI1 Antibody (27E8), DyLight 650, Novus Biologicals™

Product Code. 18423718 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423718 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423718 Supplier Novus Biologicals Supplier No. NBP222614C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ADI1 Monoclonal antibody specifically detects ADI1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADI1
Applications Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 27E8
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias 1,2-dihydroxy-3-keto-5-methylthiopentene dioxygenase, Acireductone dioxygenase (Ni(2+)-requiring), acireductone dioxygenase 1, APL1, ARDsubmergence induced protein 2, EC 1.13.11.53, FLJ10913, HMFT1638, membrane-type 1 matrix metalloproteinase cytoplasmic tail binding protein-1, Membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1, MTCBP1, MTCBP-1, Ni-ARD, SIPLMT1-MMP cytoplasmic tail-binding protein-1, Submergence-induced protein 2 homolog
Host Species Mouse
Immunogen Recombinant human ADI1 (1-179aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55256
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.