missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Adhesive spray, 3M Neoprene contact spray 80, 500mL
Click to view available options
Quantity:
500 mL
Description
All round adhesive for most plastics, laminates and wood products. Very strong with a 400 psi shear strength.
Specifications
Specifications
| Quantity | 500 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction