missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Adhesive Labels

Product Code. 15895517
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Display Text:
Chloroform
Dist. Water
Ethanol
Ethyl Acetate
Methanol
Methylene Cloride
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Display Text unitSize
15895517 Black Chloroform Pack of 50
15835517 Blue Dist. Water Pack of 50
15885517 Green Ethanol Pack of 50
15875517 Orange Ethyl Acetate Pack of 50
15855517 Black Methanol Pack of 50
15805527 Black Methylene Cloride Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15895517 Supplier Kartell™ Supplier No. 0501600
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Transparent, punched labels for use with wash bottles and plastic or glass reagent bottles

  • For use with wash bottles and plastic or glass reagent bottles, as recommended by the NFPA (National Fire Protection Agency)
  • Complete with risk assessment numbers, recommendations for use and special notices shown in reagent catalogues

Specifications

Description Transparent with colored text
Color Black
Display Text Chloroform
Quantity 50/Pk.
Dimensions (L x W) 130 x 35 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.