missing translation for 'onlineSavingsMsg'
Learn More

Agar Scientific Adhesive Carbon Tape 8mm x 20m

Brand:  Agar Scientific AGG3939

Encompass

Adhesive Carbon Tape 8mm x 20m

Product Code. 16658512

  • £45.48 / Pack of 1
Only null left
Estimated Shipment: 01-05-2026
Add to basket
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.