missing translation for 'onlineSavingsMsg'
Learn More

Color Coded High Resolution pH Buffer, Reagecon™
BIOPHARMA

Product Code. 11758465 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
pH:
4
4.00
7
7.00
10.00
Unit Size:
500mL
Each
Product Code. Quantity pH unitSize
11758465 500 mL 4.00 500mL
11778465 500 mL 10.00 500mL
11494599 500 mL 4 500mL
11404609 500 mL 7 500mL
15214643 500 mL 7 Each
11768465 500 mL 7.00 500mL
6 options
This item is not returnable. View return policy

Product Code. 11758465

Brand: Reagecon 104000C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use color coded pH buffers for ease of use. Reagecon™ Color Coded High Resolution pH Buffers are tested at either 20°C or 25°C and are certified and NIST traceable.

  • Colored pH Buffer Solutions
  • High Resolution
  • Tested at 20°C or 25°C 
  • Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
  • NIST traceable
  • Presented in various pack sizes

Specifications

Melting Point 0°C
Color Red
pH 4.00
Physical Form Liquid
Quantity 500 mL
Odor Characteristic
Solubility Information Fully miscible in water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.