missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADH7 Polyclonal Antibody
ADH7 Polyclonal Antibody
Brand: Thermo Scientific PA5104370
This item is not returnable.
View return policy
Description
ADH7 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| ADH7 | |
| Polyclonal | |
| Unconjugated | |
| ADH7 | |
| Adh3; Adh-3; Adh-3e; Adh3-e; Adh-3t; Adh3-t; Adh4; Adh7; ADH-C2; Adt-1; AI325182; alcohol dehydrogenase 3 complex; alcohol dehydrogenase 3 temporal; alcohol dehydrogenase 4; Alcohol dehydrogenase 7; alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide; alcohol dehydrogenase class 4 mu/sigma chain; Alcohol dehydrogenase class IV mu/sigma chain; alcohol dehydrogenase VII; alcohol dehydrogenase-7; All-trans-retinol dehydrogenase [NAD(+)] ADH7; class IV alcohol dehydrogenase, mu or sigma subunit; class IV sigma-1 alcohol dehydrogenase; class IV sigmasigma alcohol dehydrogenase; gastric alcohol dehydrogenase; IV ADH; Omega-hydroxydecanoate dehydrogenase ADH7; ret; Retinol dehydrogenase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 131, 171178 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P40394, P41682 | |
| ADH7 | |
| A synthesized peptide derived from human ADH7(Accession P40394), corresponding to amino acid residues D235-E285. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction