missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADH5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576022
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. The encoded protein forms a homodimer. It has virtually no activity for ethanol oxidation, but exhibits high activity for oxidation of long-chain primary alcohols and for oxidation of S-hydroxymethyl-glutathione, a spontaneous adduct between formaldehyde and glutathione. This enzyme is an important component of cellular metabolism for the elimination of formaldehyde, a potent irritant and sensitizing agent that causes lacrymation, rhinitis, pharyngitis, and contact dermatitis. The human genome contains several non-transcribed pseudogenes related to this gene.
Specifications
| ADH5 | |
| Polyclonal | |
| Unconjugated | |
| ADH5 | |
| ADH1C; Adh2; Adh-2; ADH3; ADH-3; ADH5; Adh-5; ADH-B2; ADHX; alcohol dehydrogenase (class III), chi polypeptide; alcohol dehydrogenase 1C (class I), gamma polypeptide; alcohol dehydrogenase 2; Alcohol dehydrogenase 5; alcohol dehydrogenase 5 (class III), chi polypeptide; alcohol dehydrogenase B2; Alcohol dehydrogenase class chi chain; alcohol dehydrogenase class-3; Alcohol dehydrogenase class-III; class III alcohol dehydrogenase; class III alcohol dehydrogenase, chi subunit; epididymis secretory sperm binding protein Li 60p; FALDH; FDH; formaldehyde dehydrogenase; glutathione-dependent formaldehyde dehydrogenase; GSH-FDH; GSNOR; HEL-S-60p; S-(hydroxymethyl)glutathione dehydrogenase; S-nitrosoglutathione reductase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11532, 128 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P11766, P28474 | |
| ADH5 | |
| Recombinant full length Human ADH5. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction